Learn More
ABEBA™ Uni6 White Occupational Sneakers O2
ABEBA™ Uni6 White Occupational Sneakers O2 made of tear and water resistant fabric with slip resistant PU/TPU outsole.
1090.00 NOK
Specifications
Certifications/Compliance | CE EN ISO 20347 :2012, O2, FO, SRC, ESD |
---|---|
Closure Type | Laces |
Color | White |
Eyelets Material | Stainless steel |
Liner Material | Breathable silver ions lining |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16338101
|
ABEBA™
36729/35 |
35 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16348101
|
ABEBA™
36729/36 |
36 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16358101
|
ABEBA™
36729/37 |
37 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16368101
|
ABEBA™
36729/38 |
38 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16378101
|
ABEBA™
36729/39 |
39 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16388101
|
ABEBA™
36729/40 |
40 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16398101
|
ABEBA™
36729/41 |
41 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16308111
|
ABEBA™
36729/42 |
42 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16318111
|
ABEBA™
36729/43 |
43 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16328111
|
ABEBA™
36729/44 |
44 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16338111
|
ABEBA™
36729/45 |
45 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16348111
|
ABEBA™
36729/46 |
46 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16358111
|
ABEBA™
36729/47 |
47 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
16368111
|
ABEBA™
36729/48 |
48 |
1090.00 NOK
1 pair |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
Description
- Lightweight, supple, water resistant
- Comply to EN ISO 20347 : 2012, O2, SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant PU/TPU outsole
- Good cushioning
- Energy absorption in heel area
- Tear and water resistant fabrics
- Easy fastening with laces
- Breathable and antibacterial lining with silver ions
- Sanitized treatment
- Removable Active Comfort insole
Specifications
CE EN ISO 20347 :2012, O2, FO, SRC, ESD | |
White | |
Breathable silver ions lining | |
Slip and abrasion resistant, ESD dissipative, shock absorbent | |
Trainer | |
EN ISO 20347 : 2012; DIN EN 61340 | |
Yes |
Laces | |
Stainless steel | |
Textile | |
Polyurethane, Thermoplastic Polyurethane | |
1 Pair | |
Water |
Product Title
ABEBA™ Uni6 White Occupational Sneakers O2