Learn More
Abeba™ UNI6 36761 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Brand:  Abeba™ 36761/40
 View more versions of this product
Product Code. 15660558
  • 1325.00 NOK / 1 pair
Only null left
Estimated Shipment: 08-08-2025
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRC
  • Functional leather with honeycomb pattern
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • Water-repellent
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRC
Leather
Slip-resistant
36761
Black
40
Polyurethane
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Abeba™ UNI6 36761 Shoes > Size: 40