Learn More
OHAUS™ Uni Block
Hot plates and stirrers accessory
Brand:  OHAUS™ 30500589
4515.54 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16680472
  • 4635.00 NOK / Each
Only null left
Estimated Shipment: 14-08-2025
Description
Holds test tubes or vials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mm Test Tubes
Uni Block
Metal
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OHAUS™ Uni Block > 25 mm Test Tubes