Learn More
OHAUS™ Uni Block
Hot plates and stirrers accessory
Brand: OHAUS™ 30500586
4515.54 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Holds test tubes or vials
Specifications
12 mm Test Tubes | |
Uni Block | |
Metal |
Product Content Correction
Product Title
OHAUS™ Uni Block > 12 mm Test Tubes