Learn More
OHAUS™ Uni Block
Hot plates and stirrers accessory
Brand:  OHAUS™ 30500585
4012.86 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16640472
  • 4120.00 NOK / Each
Only null left
In Stock
Description
Holds test tubes or vials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
28 mm Vials
Uni Block
Metal
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OHAUS™ Uni Block > 28 mm Vials