Learn More
Hughes Unheated Emergency Safety Shower with Eye/Face Wash
Floor mounted unheated emergency safety shower with stainless steel eye/face wash unit. This ANSI compliant model is ideal for use in indoor applications not subject to the possibility of freezing or overheating.
Brand:  Hughes HSHD0018GV
13019.72 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
 View more versions of this product
Product Code. 17702348
  • 13765.00 NOK / Each
Everyday Lab Essentials
Use promo code 24132 at checkout
View Available Promotions
Only null left
Estimated Shipment: 25-07-2025
Description
  • Floor mounted unheated emergency safety shower with stainless steel eye/face wash unit
  • ANSI compliant model ideal for use in indoor applications not subject to the possibility of freezing or overheating
  • Fitted with foot panel as standard
  • Delivers the ANSI recommended 76 liters per minute of potable water from the shower and 12 liters per minute to the eye/face wash
  • Available with either galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Specifications
Galvanized Steel
Floor
Safety Shower
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    From wipes to waste bins, labels to liners – save on them all.
    Promo Code: 24132
    Expires2025-08-24
    New Offer
    Shop Now
Product Content Correction
Product Title
Hughes Unheated Emergency Safety Shower with Eye/Face Wash > Safety Shower; Material: Galvanized Steel