Learn More
Haldenwanger™ Glazed Porcelain Mortar
Use these mortars for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Mortars are constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ L 55/0A/RAUH
Description
- Available in a variety of sizes
- Grinding surfaces are unglazed
- Complies with DIN VDE 0335, type C 100
Specifications
70 mL | |
63mm | |
Porcelain |
Unglazed | |
Mortar | |
40mm |
Accessories and Services
-
Haldenwanger™ Unglazed Porcelain Pestle Unglazed Porcelain Pestle, 150mm Product Code: 10042964 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
-
Haldenwanger™ Unglazed Porcelain Pestle Unglazed Porcelain Pestle, 135mm Product Code: 10287274
185.00 NOK / Each -
Haldenwanger™ Unglazed Porcelain Pestle Pestle, Grinding Surface Glazed, DIN 129... Product Code: 10656405
314.00 NOK / Each -
Haldenwanger™ Unglazed Porcelain Pestle Pestle, Grinding Surface Unglazed, DIN 1... Product Code: 12313338
191.00 NOK / Each -
Haldenwanger™ Unglazed Porcelain Pestle Pestle, Grinding Surface Unglazed, DIN 1... Product Code: 12343338 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
-
Haldenwanger™ Porcelain Pestle Pestle, Grinding Surface Glazed, DIN 129... Product Code: 12383338 This item is currently unavailable or has been discontinued.