Learn More
BICASA Undercounter Drawer Cabinet on Casters Product Code.: 30057571
Estimated Shipment: 11-08-2025
Only null left
Add to basket
BICASA Undercounter Drawer Cabinet on Casters
Product Code. 30057571
Light Gray, 600 x (500 + 20) x 630 mm, Each
Click to view available options
Color:
Anthracite
Blue
Light Gray
Red
Dimensions (D x W x H):
600 x (500 + 20) x 630 mm
600 x (500 + 20) x 760 mm
900 x (500 + 20) x 760 mm
Unit Size:
Each
Product Code. 30057571
Brand: BICASA TN.DW.0600.630
Only null left
Estimated Shipment: 11-08-2025
Add to basket
Only null left
Add to basket
Undercounter Drawer Cabinet on Casters with fire-retardant particleboard body and drawers with soft closing and single lock.
  • Undercounter cabinets on casters
  • Metal frame base with 4 swivel casters (Polyamide case and thermoplastic rubber treads), 2 with lockable with brakes - Height mm 100
  • Central locking system as standard
  • Anti-tilt safety system to open only one drawer at a time
  • Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
Specifications
Product Type Mobile Storage Cabinet
Color Light Gray
Cabinet Type Underbench cabinet
Dimensions (D x W x H) 600 x (500 + 20) x 630 mm
No. of Doors 1
No. of Drawers 3
Width (English) 23.6 in.
Width (Metric) 600 mm
With Casters Yes
Material Melamine
Product Content Correction
Product Title
BICASA Undercounter Drawer Cabinet on Casters >