Learn More
Vileda Professional™ UltraSpeed Pro MicroStrong Controlled Environments Mop
The UltraSpeed Pro MicroStrong Mop is a durable, 100% polyethersulfone microfiber looped mop for use with the UltraSpeed Pro CE System.
Brand: Vileda Professional™ 157778
This item is not returnable.
View return policy
Description
- Reusable, single-sided microfiber mop
- 40 cm including mop label with LOT code
- Usable up to ISO class 4 to 9 (GMP A/B, C, D)
- Strong and durable loop microfiber construction
- High absorbency
- Low particle shedding
Specifications
Mop Pad | |
Non-sterile | |
Reusable | |
Polyester |
White | |
Not Autoclavable | |
No |
Product Content Correction
Product Title
Vileda Professional™ UltraSpeed Pro MicroStrong Controlled Environments Mop > UltraSpeed Pro MicroStrong Controlled Environments Mop; Material: Polyester