Learn More
Sartorius Ultrafiltration Membrane Discs
For sample concentration in stirred cells
Brand: Sartorius 14629--76------D
4223.26 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.
- Wide range of MWCO
- Wide range of disc diameters
- Compatible with all stirred cells
Specifications
Membrane Filter | |
PES | |
5 kDa | |
For sample concentration in stirred cells | |
120 ÎĽm | |
76 mm |
Ultrafiltration Disc | |
Polysulfone | |
Non-sterile | |
10/Pk. | |
Membrane Filter |