Learn More
Invitrogen™ UltraComp eBeads™ Plus Compensation Beads
UltraComp eBeads Plus Compensation Beads are the second generation of UltraComp beads for use in flow cytometry.
Brand: Invitrogen™ 01-3333-41
This item is not returnable.
View return policy
Description
- Binds the spherical particles for use as a single-color compensation controls.
- Binds with mouse, rat and hamster antibodies, but also bind with rabbit and recombinant human antibodies.
- Designed for use in compensation with all fluorochromes excited by ultraviolet (355 nm) violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), and red (633-640 nm) lasers.
- Compensate in a manner similar to cells when used with Brilliant Violet 785/786-, Brilliant Violet 711-, Super Bright 780- and Super Bright 702-conjugated antibodies.
- Multispecies compatibility—bind with mouse, rat, hamster, rabbit and recombinant human antibodies
- Improved fluorophore compatibility
- Each drop of beads contains two populations: a positive population that bind with any mouse, rat, hamster, rabbit or recombinant human antibody and a negative population that will not bind with antibody.
- When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations are observed.
- This bimodal distribution can be used for single-color compensation controls to accurately setup instrumentation for multicolor flow cytometry experiments.
Specifications
Flow Cytometry | |
eBeads™ | |
Compensation Beads |
25 | |
25 tests |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Sheet: UltraComp eBeads Compensation Beads and UltraComp eBeads Plus Compensation Beads
Version: B.0
File size: 349.5 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3071389 | Certificate of Analysis | 25/10/2024 | 01-3333-41 |
2897791 | Certificate of Analysis | 08/05/2024 | 01-3333-41 |
2811998 | Certificate of Analysis | 15/03/2024 | 01-3333-41 |
2697000 | Certificate of Analysis | 29/09/2023 | 01-3333-41 |
2659309 | Certificate of Analysis | 31/05/2023 | 01-3333-41 |
Product Content Correction
Product Title
Invitrogen™ UltraComp eBeads™ Plus Compensation Beads > 25 Tests
RUO use only.