Learn More
Arctiko™ ULTF 80 Ultra Low Temperature Freezer
The ULTF range exhibits high-performance freezing with a focus on low energy consumption.
Brand:  Arctiko™ 1100001
Product Code. 11697620
  • 62498.58 NOK / Each
Description
The ULTF range exhibits high-performance freezing with a focus on low energy consumption. The range has been updated with our advanced Arctiko™ controller, which ensures that all alarm and data logging features are included as standard. The ULTF range is also produced with our original single compressor technology to ensure the lowest possible noise level.
  • Single compressor technology
  • Low energy consumption
  • Low noise level (50 dBa)
  • Low heat dissipation
  • Sub lids
  • Key lock on handle
  • Digital display
  • 100% HCFC and CFC free
  • Filterless construction
  • Porthole for external temperature probes
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Arctiko™ ULTF 80 Ultra Low Temperature Freezer > Capacity: 71L