missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE4B Rabbit anti-Human, Mouse, Rat, Clone: 1D7K5, Novus Biologicals™
Rabbit Monoclonal Antibody
1815.00 NOK - 4580.00 NOK
Specifications
Antigen | UBE4B |
---|---|
Clone | 1D7K5 |
Dilution | Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunohistochemistry-Paraffin 1:20-1:50 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18329486
|
Bio-Techne
NBP3-15263-20UL |
20 μg |
1815.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18334875
|
Bio-Techne
NBP3-15263-100UL |
100 μg |
4580.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBE4B Monoclonal antibody specifically detects UBE4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
UBE4B | |
Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunohistochemistry-Paraffin 1:20-1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
E4, homologous to yeast UFD2, Homozygously deleted in neuroblastoma 1, homozygously deleted in neuroblastoma-1, KIAA0684HDNB1, ubiquitin conjugation factor E4 B, Ubiquitin fusion degradation protein 2, ubiquitination factor E4B (homologous to yeast UFD2), ubiquitination factor E4B (UFD2 homolog, yeast), ubiquitin-fusion degradation protein 2, UBOX3, UFD2UFD2A | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBE4B (O95155). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
1D7K5 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10277 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title