Learn More
UBE2D3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11677-1-AP-20UL
 View more versions of this product
Product Code. 16898913
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2D3 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme functions in the ubiquitination of the tumor-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase. The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme functions in the ubiquitination of the tumor-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase. Multiple spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been determined.
Specifications
Specifications
UBE2D3
Polyclonal
Unconjugated
UBE2D3
E2(17)KB3, UBC4/5, UBC5C, UBCH5C, UBE2D3, Ubiquitin carrier protein D3, Ubiquitin protein ligase D3
Rabbit
Antigen Affinity Chromatography
RUO
66105, 7323, 81920
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.87 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P61077, P61078, P61079
UBE2D3
UBE2D3 Fusion Protein Ag2278
20 μL
Primary
Mouse, Human, Rat