Learn More
SI Analytics™ Ubbelohde Viscometer (DIN)
Ubbelohde viscometers are calibrated for manual measurements
Brand:  SI Analytics™ 285400115
Product Code. 10242071
  • 5990.00 NOK / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Description
  • Calibrated for manual measurement
  • DIN
  • Type No. 501 31
  • Capillary No. IIIa
  • Constant K = 5
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ubbelohde Viscometer