Learn More
SI Analytics™ Ubbelohde Viscometer (ASTM)
Ubbelohde viscometers are calibrated for automatic measurements
Brand: SI Analytics™ 285401255
5592.02 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Calibrated for automatic measurement
- ASTM
- Type No. 527 00
- Capillary No. 0
- Constant K = 0.001
Specifications
Ubbelohde Viscometer |
SHO014 UBBELOHDE VISCOMETER
File size: 21.3 MB
Product Content Correction
Product Title
SI Analytics™ Ubbelohde Viscometer (ASTM) > For automatic measurement