Learn More
Poulten Selfe™ Ubbelohde Suspended-Level Viscometers
Suitable for transparent liquids. With UKAS certificate of calibration
Brand: Poulten Selfe™ 1643/08
This item is not returnable.
View return policy
Description
ASTM D445-IP71, ASTM D446.Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.Specifications
Ubbelohde Suspended-Level Viscometer | |
D445-IP71, D446 | |
2C |
60 to 300 cSt | |
D446 | |
18 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Accessories and Services
Optional Accessory
-
Poulten Selfe™ Viscometer Holder VISCOMETER HOLDER,SUITABLE FOR 50MM BATH... Product Code: 13111103
1975.00 NOK / Each
Product Content Correction
Product Title
Poulten Selfe™ Ubbelohde Suspended-Level Viscometers > Size: 2C; Viscosity Range: 60 to 300mm2/s