missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UACA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25654-1-AP-150UL
This item is not returnable.
View return policy
Description
Regulates APAF1 expression and plays an important role in the regulation of stress-induced apoptosis. Promotes apoptosis by regulating three pathways, apoptosome up-regulation, LGALS3/galectin-3 down-regulation and NF-kappa-B inactivation. Regulates the redistribution of APAF1 into the nucleus after proapoptotic stress. Down-regulates the expression of LGALS3 by inhibiting NFKB1.Specifications
UACA | |
Polyclonal | |
Unconjugated | |
UACA | |
KIAA1561, NUCLING, UACA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55075, 72565 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CGB3, Q9BZF9 | |
UACA | |
UACA Fusion Protein Ag22438 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction