Learn More
DuPont™ Tyvek™ IsoClean™ Coverall with Shoe Covers
Composed of flash spun high-density polyethylene. Dupont™ Tyvek™ IsoClean™ Coverall with Shoe Covers is permeable to both air and water vapor, yet repels water-based liquids and aerosols. Offers an excellent barrier against fine particles and fibers (down to 1μ in size) and is ultra-low-linting.
Brand: DuPont™ D14248016
Description
- Coverall with hood
- White
- Clean-processed and sterile
- Stitched internal seams
- Set sleeve design with elastication at wrists, ankles and face
- Attached thumb loops
- Zipper closure
- Attached overshoes with slip-retardant soles
- Not CE certified as chemical protective clothing
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AHS2520140 | Certificate of Sterility | 12/06/2025 | D14248016 |
AHS2516128 | Certificate of Sterility | 12/06/2025 | D14248016 |
AHS2516125 | Certificate of Sterility | 12/06/2025 | D14248016 |
AHS2515153 | Certificate of Sterility | 12/06/2025 | D14248016 |
AHS2515128 | Certificate of Sterility | 12/06/2025 | D14248016 |