Learn More
DuPont™ Tyvek™ IsoClean™ Chemo Gown
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Beschrijving
- Available in sizes XS to 3X
- Increased skirt length ensures protection against frontal exposure Openness in the back closure increases wearer comfort
- Delivers an ideal balance of protection, durability and comfort
- Made of high density polyethylene using a patented flash spinning process
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based liquid splash
- Manufactured in a controlled environment
Specificatie
Specificatie
Size | X-Small |
Product Type | Controlled Environments Gown |
Material | Polyethylene |
Color | White |
Closure Type | Hook and Loop |
Disposable | Single-use |
Sterility | Non-sterile |
Cuff Style | Knit |
Latex Free | Yes |
Sleeve Style | Long Sleeve |
Meer weergeven |
Documentatie
Scientific Resources
Brochures
SDS_17876865_EN.pdf
File size: 472.4 kB
CE_17876865_EN.pdf
File size: 1.1 MB
Product Content Correction
Product Title
DuPont™ Tyvek™ IsoClean™ Chemo Gown >