Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Bag
GREENER_CHOICE
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. These filling needle bags stay in place via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically removed with forceps after the filling needles are set in place, without having to cut, or remove particulate generating autoclave tape.
Brand:  Keystone Cleanroom Products BGT2A0211D
 View more versions of this product
Product Code. 17737877
  • 54690.00 NOK / Pack of 500
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Description
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Autoclavable
651 mL
Drawstring
Single-use
Pharmaceutical Equipment
Non-sterile
<250°F
11 x 2 in.
28 cm
5 cm
22 oz.
COC
White
Steam Sterilization
Tyvek
Steam
<125°C
11 in.
2 in.