Learn More
Saint Gobain Life Sciences Tygon™ F-4040-A Tubing
Tygon™ F4040-A Tubing is tailored for petro-chemicals. This tubing resists swelling and cracking, and is UV and ozone resistant.
Brand: Saint Gobain Life Sciences T4024-43
This item is not returnable.
View return policy
Description
- Easily identifiable yellow, flexible tubing for transportation of petroleum, heating oils, kerosene and glycol based coolants.
- Resists swelling and brittleness, and the minimum extractibility safeguards the fluids being handled.
- Not for use with strong acids or alkalis.
Specifications
Tubing | |
19 mm | |
1 in. | |
1/8 in. | |
Transporting Fuel | |
Flexible |
0.75 in. | |
Tygon | |
25.4 mm | |
3.2 mm | |
Yellow | |
1.38 bar |