missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences™ Tygon S3™ Twin Tube A24-C

Black Vacuum Tubing for use with milking equipment.

Brand:  Saint Gobain Life Sciences™ ACR02917

Product Code. 17903591

  • 3289.46 NOK / 30m

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ideal for raw milk transport lines and dairy processing
  • Smooth inner surface to help prevent blockages that can restrict airflow, reducing the need for maintenance
  • Phthalate-free air transport tubing is designed to work with our clear, flexible Tygon™ M-34-R tubing that eliminates bacterial growth within milk transport lines
  • When combined, these two products create a vacuum of air that produces a continuous, reliable flow during the milking process
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tubing
0.283 in.
Thermoplastic
13.5 mm
30 m
177 lbf/in.
56 (Shore A)
25 psi
165°F
Not Autoclavable
7.2 mm
0.531 in.
98.4 in.
Black Vacuum Tubing for Milking Equipment
Black
28 mm bend radius
29.9 in. Hg (at 73°F)
74°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers