Learn More
Thermo Scientific™ TX-750 Round Bucket Adapters
Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.
Brand: Thermo Scientific™ 75003723
8253.63 NOK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Specifications
Round Bucket Adapter | |
5/7 mL Blood Collection Tube | |
5/7 mL Blood Collection Tube | |
Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31