Learn More
Lenz Laborglasintrumente™ Two-Neck Round-Bottom Flasks
Distillation flasks with parallel side neck and conical ground joints
Brand: Lenz Laborglasintrumente™ 03204149
This item is not returnable.
View return policy
Description
- With inscription label
- Material: DURAN™
Specifications
Center neck - NS 29/32, Side Neck - NS 14/23 | |
Borosilicate Glass (3.3) | |
85 mm |
250 mL | |
Distillation |