Learn More
Labplas Twirl'EM™ Clear Polyethylene Sampling Bags
Collect solid, semi-solid or liquid samples in the laboratory using Twirl'EM™ Clear Polyethylene Sampling Bags. The bags feature a twirl round metal wire closure system with safety tabs to provide a leakproof seal.
Brand:  Labplas EFL-1012
 View more versions of this product
Product Code. 11582773
  • 6300.00 NOK / Pack of 1000
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Description
  • Sterile
  • Clear polyethylene
  • Without side seals, opening at width
  • Pull tabs to open bag without touching edges
  • Twirl round metal wire closure system with safety tabs
Specifications
Specifications
2250 mL
Polyethylene
1000 Pk.
Clear
Sterile
SDS
Documents Certificates
Product Title
Labplas Twirl'EM™ Clear Polyethylene Sampling Bags > Capacity: 2250mL