Learn More
Labplas Twirl'EM™ Clear Polyethylene Sampling Bags
Collect solid, semi-solid or liquid samples in the laboratory using Twirl'EM™ Clear Polyethylene Sampling Bags. The bags feature a twirl round metal wire closure system with safety tabs to provide a leakproof seal.
Brand: Labplas EPL-3070-FS-1_OLD
Description
- Sterile
- Clear polyethylene
- Without side seals, opening at width
- Pull tabs to open bag without touching edges
- Twirl round metal wire closure system with safety tabs
Specifications
150 mL | |
Polyethylene | |
1000 Pk. |
Clear | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Product Title
Labplas Twirl'EM™ Clear Polyethylene Sampling Bags > Capacity: 150mL