Learn More
Brannan™ Twaddell Hydrometers
Shop loaded Twaddell hydrometers for specific gravity range 24 to 48g/mL
Brand:  Brannan™ 63/100/0
 View more versions of this product
Product Code. 12859207
  • 233.00 NOK / Each
Only null left
Estimated Shipment: 09-07-2025
Description
For 0 to 24 Twaddell range subdivided 1.0TW. 158mm Shot loaded, scale subdivision 1.0TW, accuracy ±1.0TW, 0.5TW ±0.5TW at 60/60°F. Calibrated at 15.6°C.
Specifications
Specifications
1Pack
±1Tw
250 mm
Twaddle Hydrometer
1.0TW
48 to 74 g/mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brannan™ Twaddell Hydrometers > Specific Gravity Range: 48 to 74g/mL