Learn More
Brannan™ Twaddell Hydrometers
Shop loaded Twaddell hydrometers for specific gravity range 24 to 48g/mL
Brand:  Brannan™ 63/099/9
 View more versions of this product
Product Code. 12849207
  • 218.00 NOK / Each
Only null left
Estimated Shipment: 08-07-2025
Description
For 0 to 24 Twaddell range subdivided 1.0TW. 158mm Shot loaded, scale subdivision 1.0TW, accuracy ±1.0TW, 0.5TW ±0.5TW at 60/60°F. Calibrated at 15.6°C.
Specifications
Specifications
1Pack
±1Tw
250 mm
Twaddle Hydrometer
1.0TW
24 to 48 g/mL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brannan™ Twaddell Hydrometers > Specific Gravity Range: 24 to 48g/mL