Learn More
Thermo Scientific™ Turbidity Standards for Orion™ AQ3010 Turbidity Meter
Replacement Turbidity Standards for the Orion AQ3010 Turbidity meter.
Brand:  Thermo Scientific™ AC301S
3867.72 NOK valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 12013273
  • 4197.34 NOK / Pack of 4
Explore more special offers
Description
Description
Turbidity Standards are for use with the Orion™ AQ3010 Turbidity Meter (Cat. No. AQ3010). These replacement calibration standards kit are polymer-based primary turbidity standards and includes 800, 100, 20 and 0.02 NTU in 60mL plastic bottles.
  • For use with: Orion AQUAfast AQ3010
  • Standards Set has 60 mL bottle each of 0.02, 20.0, 100, and 800 NTU 100
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibration Standards
Orion AQ3010 Turbidity Meter
Replacement calibration standards kit, includes 800, 100, 20.0 and 0.02 NTU standards in plastic bottles
800, 100, 20.0 and 0.02 NTU standards in 60mL bottles
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number035-01 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog NumberAC301S
Lot Number474-01 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog NumberAC301S
Lot Number434-01 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog NumberAC301S
Lot Number374-01 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog NumberAC301S
Lot Number274-01 Certificate TypeCertificate of Analysis Date05/07/2024 Catalog NumberAC301S
Product Title
Thermo Scientific™ Turbidity Standards for Orion™ AQ3010 Turbidity Meter >