Learn More
WTW™ Turb™ Series 430 Portable Turbidity Meters
With an accuracy of 0.01 NTU or ±2% of the measured value; these portable turbidity meters are ideal for all applications including drinking water testing, environmental monitoring, laboratory reference measurements and process control.
Brand: WTW™ 600325
26848.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Portable turbidity meter with automatic calibration type
- Accuracy of 0.01 NTU or ±2% of the measured value
- Includes single instrument with AMCO Clear KalKit (0.02-10-1000 NTU), 5 empty vials, 4 Mignon batteries, optical cleaning tissues
- Used for all applications including drinking water testing, environmental monitoring, laboratory reference measurements and process control
- Resolution of 0.01 from 0.00 to 9.99, 0.1 from 10 to 99.90, 1 from 100 to 1100
Specifications
Portable Turbidity Meter | |
LCD | |
Turb™ 430 T Portable turbidity meter | |
Single instrument with AMCO Clear KalKit (0.02-10-1000 NTU), 5 empty vials, 4 Mignon batteries, optical cleaning tissues. (Optional LabStation with PC-Software and rech. battery (251-301) | |
Used for all applications including drinking water testing, environmental monitoring, laboratory reference measurements and process control | |
0.01 NTU or ±2% of the measured value | |
117 mm | |
4 x AA | |
0°C to 50°C | |
2 Years |
Tungsten | |
7 sec. | |
Automatic | |
20 mL | |
IP67 | |
0.01 from 0.00 to 9.99, 0.1 from 10 to 99.90, 1 from 100 to 1100 | |
86 mm | |
CE, US EPA 180.1 | |
0.6 kg |