Learn More
Palintest™ Tubetest COD Reagent Tube with Mercury For Moderate Chloride Sample
Ideal for use in standard procedures which require the suppression of interference due to moderate levels of chloride.
951.88 NOK - 1033.50 NOK
Specifications
For Use With (Application) | Moderate Chloride Samples Such as Chemical Oxygen Demand (COD) |
---|---|
For Use With (Equipment) | Photometer or UV-Vis Spectrophotometer |
Includes | Chemical Oxygen Demand (COD) |
No. of Tests | 25 |
Type | COD Reagent Tube |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
11938927
|
Palintest™
PL460 |
0 to 150mg/L |
951.88 NOK
Pack of 24 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11956473
|
Palintest™
PL466 |
0 to 20000mg/L |
1033.50 NOK
Pack of 24 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The mercury free formulation is recommended for general use
- The mercuric sulfate formulation (M) is provided for use in standard procedures which require the suppression of interference due to moderate levels of chloride
- The high chloride formulation (2M) provides much higher levels of chloride suppression and is suitable for working with samples with very high chloride levels
- The new M/C formulation with mercuric sulfate is specifically for use in non-Palintest™ instruments
Specifications
Moderate Chloride Samples Such as Chemical Oxygen Demand (COD) | |
Chemical Oxygen Demand (COD) | |
COD Reagent Tube |
Photometer or UV-Vis Spectrophotometer | |
25 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- COD 150/M
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P314-Get medical advice/attention if you feel unwell.
- P321-Specific treatment (see on this label).
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P363-Wash contaminated clothing before reuse.
- P391-Collect spillage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, mercuric sulphate, silver sulphate, potassium dichromate