Learn More
TST™ Bowie Dick™ Test Pack
Test the steam penetration and air removal capability of a vacuum sterilizer with this test pack. The TST™ Bowie Dick Test Pack is a unique combination of advance Browne intelligent ink technology and the original Bowie Dick concept.
Brand: Steris Corporation 2352AB
This item is not returnable.
View return policy
Description
- Single-use test pack
- A successful test confirms that steam penetration into a test pack is rapid and even and that air and other non-condensable gases have been effectively removed
- The chemical indicator sheet at the center of the pack shows a defined color change from yellow to dark blue/purple when exposed to a specific combination of time, temperature and steam
- Conforms to ISO 11140-4 Class 2
Specifications
Autoclavable | |
For large porous load sterilizers | |
TST Bowie Dick Test Pack | |
134 to 137˚C for up to 3.5 min., Conforms to ISO 11140-4(3) Class 2 | |
Yes | |
Steam |
ISO 11140-4(3) Class 2 | |
Autoclave | |
134°C to 137°C | |
100 Pk. | |
3.5 min. |
Product Content Correction
Product Title
TST™ Bowie Dick™ Test Pack > Sterilizer range: 134deg.C to 137deg.C for up to 3.5 minutes