Learn More
TSPAN15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10460-1-AP-20UL
 View more versions of this product
Product Code. 16806863
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
TSPAN15 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene.
Specifications
Specifications
TSPAN15
Polyclonal
Unconjugated
TSPAN15
2700063A19Rik, NET 7, NET7, Tetraspan NET 7, tetraspanin 15, TM4SF15, Tspan 15, TSPAN15
Rabbit
Antigen Affinity Chromatography
RUO
23555
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95858
TSPAN15
TSPAN15 Fusion Protein Ag0710