missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55455-1-AP-150UL
This item is not returnable.
View return policy
Description
TRPM6 is predomitly expressed in the kidney and colon, and encodes a protein containing an ion channel domain and a protein kinase domain. TRPM6 is crucial for magnesium homeostasis, and plays an essential role in epithelial magnesium transport and in the active magnesium absorption in the gut and kidney. Further, TRPM6 is an essential ion channel and serine/threonine-protein kinase, and is crucial for magnesium homeostasis. TRPM6 has an important role in epithelial magnesium transport and in the active magnesium absorption in the gut and kidney. Isoforms of the type M6-kinase lack the ion channel region. Mutations in TRPM6 are associated with hypomagnesemia with secondary hypocalcemia. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Specifications
TRPM6 | |
Polyclonal | |
Unconjugated | |
TRPM6 | |
CHAK2, Channel kinase 2, HMGX, HOMG, HOMG1, HSH, TRPM6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140803 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BX84 | |
TRPM6 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction