Learn More
Troemner Replacement Wall Bracket (Labjaws)
Secure gas cylinders to increase safety with this Wall Bracket (Labjaws).
Brand:  Troemner 08129-64
1353.75 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Product Code. 15220289
  • 1425.00 NOK / Each
Everyday Lab Essentials
Use promo code 24132 at checkout
View Available Promotions
Description
  • 2 year warranty.
Specifications
Specifications
Nylon, Nickel Plated Zinc
Grip Range: 4 to 14 in.
Replacement Wall Bracket (Labjaws)
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Troemner Replacement Wall Bracket (Labjaws) > Replacement Wall Bracket (Labjaws); Grip Range: 4 to 14 in.