Learn More
TrkA Monoclonal Antibody (6B2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515509
3174.50 NOK valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
MA5-15509 targets TrkA in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15509 immunogen is purified recombinant extracellular fragment of human TrkA (aa33-423) fused with hIgGFc tag expressed in HEK293 cell line. MA5-15509 detects TrkA which has a predicted molecular weight of approximately 87.4kDa.
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
Specifications
TrkA | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P04629 | |
Ntrk1 | |
Purified recombinant extracellular fragment of human TrkA (aa33-423) fused with hIgGFc tag expressed in HEK293 cell line. | |
RUO | |
4914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
6B2 | |
Unconjugated | |
Ntrk1 | |
C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; Trka; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.