missing translation for 'onlineSavingsMsg'
Learn More

TRIS buffer, Tris-(hydroxymethyl)-aminomethane, molecular biology grade, Scharlab

Product Code. 11382605
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 77-86-1
Molecular Formula C4H11NO3
Quantity 1 kg
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES NC(CO)(CO)CO
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 121.14
Grade Molecular Biology
compliance-icons
Product Identifier
  • Tris-(hydroxymethyl)-aminomethane, molecular biology grade
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P362-Take off contaminated clothing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'