Learn More
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride, Thermo Scientific Chemicals
Redox indicator, oxygen sensor
3630.00 NOK - 12190.00 NOK
Chemical Identifiers
CAS | 36309-88-3 |
---|---|
Molecular Formula | C72H48Cl2N6Ru |
Molecular Weight (g/mol) | 1169.188 |
MDL Number | MFCD03095387 |
InChI Key | SKZWFYFFTOHWQP-UHFFFAOYSA-L |
Synonym | bathophenanthroline ruthenium chloride, ru ddp, tris-bathophenanthroline ruthenium ii chloride, dichlororuthenium-4,7-diphenyl-1,10-phenanthroline 1/3, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride, tris 4,7-diphenyl-1,10-phenanthroline ; dichlororuthenium, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, for fluorescence |
PubChem CID | 15787787 |
IUPAC Name | dichlororuthenium;4,7-diphenyl-1,10-phenanthroline |
SMILES | C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.Cl[Ru]Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309608
|
Thermo Scientific Alfa Aesar
044123.02 |
0.5 g |
3630.00 NOK
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319608
|
Thermo Scientific Alfa Aesar
044123.04 |
2 g |
12190.00 NOK
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride is used as a probe for luminescent detection and quantitation of oxygen. It is also used for fiber optic sensors. Further, it is used in studies on oxygen in skin and skin tumors as well as measurement of oxygen flux through the skin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
36309-88-3 | |
1169.188 | |
SKZWFYFFTOHWQP-UHFFFAOYSA-L | |
15787787 | |
C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.Cl[Ru]Cl |
C72H48Cl2N6Ru | |
MFCD03095387 | |
bathophenanthroline ruthenium chloride, ru ddp, tris-bathophenanthroline ruthenium ii chloride, dichlororuthenium-4,7-diphenyl-1,10-phenanthroline 1/3, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride, tris 4,7-diphenyl-1,10-phenanthroline ; dichlororuthenium, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, for fluorescence | |
dichlororuthenium;4,7-diphenyl-1,10-phenanthroline |
Specifications
36309-88-3 | |
MFCD03095387 | |
Soluble in alcohol and dimethylformamide. Insoluble in water. | |
C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.Cl[Ru]Cl | |
1169.188 | |
1169.2 | |
Tris (4,7-diphenyl-1,10-phenanthroline)ruthenium (II) dichloride |
C72H48Cl2N6Ru | |
bathophenanthroline ruthenium chloride, ru ddp, tris-bathophenanthroline ruthenium ii chloride, dichlororuthenium-4,7-diphenyl-1,10-phenanthroline 1/3, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride, tris 4,7-diphenyl-1,10-phenanthroline ; dichlororuthenium, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, for fluorescence | |
SKZWFYFFTOHWQP-UHFFFAOYSA-L | |
dichlororuthenium;4,7-diphenyl-1,10-phenanthroline | |
15787787 | |
Powder |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.