missing translation for 'onlineSavingsMsg'
Learn More
Trimethyltin chloride, 99% Product Code.: 10225640

Trimethyltin chloride, 99%

Product Code. p-3737699 Shop All Thermo Scientific Chemicals Products
10 g, Glass bottle, 10g
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10225640

Brand: Thermo Scientific Acros 163990100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1066-45-1 | C3H11ClSn | 201.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-45-1
Molecular Formula C3H11ClSn
Molecular Weight (g/mol) 201.28
MDL Number MFCD00000520
InChI Key FVFGWISLDLUGHX-UHFFFAOYSA-N
Synonym trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
PubChem CID 14016
IUPAC Name chloro(trimethyl)stannane
SMILES Cl.C[SnH](C)C

Specifications

Melting Point 37.0°C to 39.0°C
Color White
Boiling Point 148.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SnCl
Quantity 10 g
Fieser 17,388; 03,19; 04,27; 05,26; 06,41; 07,18; 13,239
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,organic solvents
Formula Weight 199.25
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Trimethyltin chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyltin chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethyltin chloride, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.