missing translation for 'onlineSavingsMsg'
Learn More
Trimethylacetaldehyde, 95%, Thermo Scientific Chemicals Product Code.: 11417257

Trimethylacetaldehyde, 95%, Thermo Scientific Chemicals

Product Code. 11417257
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11417257

Brand: Thermo Scientific Alfa Aesar A15013.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 630-19-3 | C5H10O | 86.13 g/mol

Trimethylacetaldehyde is used as a stereoselective synthesis application. It is also used as a building block used in aldol condensation reactions. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetaldehyde is used as a stereoselective synthesis application. It is also used as a building block used in aldol condensation reactions. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is virtually insoluble in water but soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from oxidizing agents, strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-19-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006962
InChI Key FJJYHTVHBVXEEQ-UHFFFAOYSA-N
Synonym trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehydeShow MoreShow Less
PubChem CID 12417
IUPAC Name 2,2-dimethylpropanal
SMILES CC(C)(C)C=O

Specifications

Melting Point 6°C
Density 0.775
Boiling Point 74°C to 76°C
Flash Point −15°C (5°F)
Odor Pleasant
Linear Formula (CH3)3CCHO
Refractive Index 1.3794
Quantity 100 g
UN Number UN1989
Beilstein 506060
Sensitivity Air sensitive
Solubility Information It is virtually insoluble in water but soluble in chloroform.
Formula Weight 86.13
Percent Purity 96%
Chemical Name or Material Trimethylacetaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0471836 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberA15013.06,A15013.14,A15013.22
Lot NumberA0464314 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberA15013.06,A15013.14,A15013.22
Lot NumberA0461260 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog NumberA15013.06,A15013.14,A15013.22
Lot NumberA0444238 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberA15013.06,A15013.14,A15013.22
Lot Number10210459 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15013.06,A15013.14,A15013.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethylacetaldehyde, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.