missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10851-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nucleus and the cytoplasm. Its function has not been identified.Specifications
TRIM11 | |
Polyclonal | |
Unconjugated | |
TRIM11 | |
BIA1, Protein BIA1, RING finger protein 92, RNF92, TRIM11, tripartite motif containing 11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
81559 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96F44 | |
TRIM11 | |
TRIM11 Fusion Protein Ag1308 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction