missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol diacetate, 98%, Thermo Scientific™

Product Code. 11303376
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-21-7
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.248
MDL Number MFCD00026213
InChI Key OVOUKWFJRHALDD-UHFFFAOYSA-N
Synonym triethylene glycol diacetate, tdac, triglycol diacetate, triethylene glycol, diacetate, triglycol, diacetate, acetic acid, triethylene glycol diester, triethyleneglycoldiacetate, 2,2'-ethylenedioxy di ethyl acetate, ethanol, 2,2'-ethylenedioxydi-, diacetate, ethanol, 2,2'-1,2-ethanediylbis oxy bis-, diacetate
PubChem CID 8098
IUPAC Name 2-[2-(2-acetyloxyethoxy)ethoxy]ethyl acetate
SMILES CC(=O)OCCOCCOCCOC(=O)C

Specifications

Melting Point -50°C
Density 1.12
Boiling Point 286°C
Flash Point 163°C (325°F)
Refractive Index 1.44
Quantity 1000 g
Beilstein 1789453
Formula Weight 234.25
Percent Purity 98%
Chemical Name or Material Triethylene glycol diacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'