Learn More
Triethanolamine, 97%, Thermo Scientific Chemicals Product Code.: 10657354
Triethanolamine, 97%, Thermo Scientific Chemicals
Product Code. 10657354 Shop All Thermo Scientific Chemicals Products
25 L, 25 L
Click to view available options
Quantity:
25 L
Unit Size:
25 L
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10657354
Brand: Thermo Scientific Acros 139560251
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102-71-6
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym trolamine, 2,2',2-nitrilotriethanol, thiofaco t-35, sterolamide, trihydroxytriethylamine, nitrilotriethanol, tris 2-hydroxyethyl amine, triethylolamine, daltogen, triethanolamine
PubChem CID 7618
ChEBI CHEBI:28621
Specifications
Decomposition Information >325°C
Melting Point 21.0°C
Density 1.1245 g/mL
Boiling Point 360.0°C
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H15NO3
Linear Formula (HOCH2CH2)3N
Refractive Index 1.4840 to 1.4860
Quantity 25 L
Fieser 01,1196
Merck Index 15, 9830
Specific Gravity 1.1245
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohol,methanol and acetone,soluble in chloroform,slichtly soluble in benzene and ether,soluble 4.2% in benzene; 1.6% in ether; 0.4% in,carbon tetrachloride and < 0.1% in n-he
SMILES OCCN(CCO)CCO
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Viscosity 600 mPa.s (25°C)
Formula Weight 149.19
Vapor Pressure <0.01 mmHg (at 20°C)
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material Triethanolamine, 97%
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0471278 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code139560251,139561000
Lot NumberA0448795 Certificate TypeCertificate of Analysis Date25/01/2025 Product Code139560025,139561000,139560251,139560010
Lot NumberA0448795 Certificate TypeCertificate of Analysis Date05/01/2024 Product Code139560025,139560251,139561000,139560010
Lot NumberB0540414 Certificate TypeCertificate of Analysis Date20/10/2023 Product Code139560025,139560251,139561000,139560010
Lot NumberA0432956 Certificate TypeCertificate of Analysis Date25/08/2023 Product Code139560025,139560251,139561000,139560010
Product Title
Triethanolamine, 97%, Thermo Scientific Chemicals >