missing translation for 'onlineSavingsMsg'
Learn More
Trichloroacetic Acid, Reagent Grade, ACS, Scharlab Product Code.: 11322605
compliance-icons compliance-icons

Trichloroacetic Acid, Reagent Grade, ACS, Scharlab

Product Code. 11322605
100 g, HDPE bottle, 100g
Click to view available options
Quantity:
100 g
Packaging:
HDPE bottle
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11322605

Brand: Scharlab AC31320100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Complies with American Chemical Society specifications. Trichloroacetic Acid, Reagent Grade, ACS, Scharlab is packed in an HDPE container for great resistance and durability to ensure product safety and quality.

Reagent grade chemicals are the most frequently used grade for laboratory analytical work. They correspond to a grade of high quality chemicals for laboratory and specialized industrial use.

Specifications

CAS 76-03-9
Melting Point 54°C to 58°C
Boiling Point 197°C
Color white
Molecular Formula C2HCl3O2
Linear Formula CCl3COOH
UN Number 1839
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
Solubility Information Soluble in water
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES C(=O)(C(Cl)(Cl)Cl)O
IUPAC Name 2,2,2-trichloroacetic acid
Molecular Weight (g/mol) 163.378
PubChem CID 6421
ChEBI CHEBI:30956
Formula Weight 163.39g/mol
Percent Purity 99.5% (min.)
Grade ACS
Quantity 100 g
Packaging HDPE bottle
Physical Form Solid
Residue after Ignition Max. 0.03%
Vapor Pressure 1 hPa
Chemical Name or Material Trichloroacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trichloroacetic acid, reagent grade, ACS
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trichloroacetic Acid, Reagent Grade, ACS, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.