missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylamine, ≥98.5%, Honeywell™
≥98.5%
325.05 NOK - 1878.00 NOK
Chemical Identifiers
CAS | 102-82-9 |
---|---|
Molecular Formula | C12H27N |
Molecular Weight (g/mol) | 185.355 |
MDL Number | MFCD00009431 |
InChI Key | IMFACGCPASFAPR-UHFFFAOYSA-N |
Synonym | tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7 |
PubChem CID | 7622 |
ChEBI | CHEBI:38905 |
IUPAC Name | N,N-dibutylbutan-1-amine |
SMILES | CCCCN(CCCC)CCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
15648410
|
Honeywell Research Chemicals
471313-25ML |
25 mL | Glass Bottle | N/A | ||||||
15625630
|
Honeywell Research Chemicals
471313-250ML |
250 mL | Glass Bottle | N/A | ||||||
15635630
|
Honeywell Research Chemicals
471313-2L |
2 L | Glass Bottle | N/A | ||||||
15615630
|
Honeywell Research Chemicals
471313-2.5L |
2.5 L | Glass Bottle | N/A | ||||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
102-82-9 | |
185.355 | |
IMFACGCPASFAPR-UHFFFAOYSA-N | |
7622 | |
N,N-dibutylbutan-1-amine |
C12H27N | |
MFCD00009431 | |
tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7 | |
CHEBI:38905 | |
CCCCN(CCCC)CCCC |
Specifications
102-82-9 | |
Colorless | |
216°C | |
Unpleasant | |
[CH3(CH2)3]3N | |
UN2542 | |
tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7 | |
CCCCN(CCCC)CCCC | |
185.355 | |
CHEBI:38905 | |
≥98.5% | |
Tributylamine |
-70°C | |
0.778g/mL at 25°C | |
63°C (145.4°F) | |
C12H27N | |
MFCD00009431 | |
1698872 | |
IMFACGCPASFAPR-UHFFFAOYSA-N | |
N,N-dibutylbutan-1-amine | |
7622 | |
185.35g/mol | |
Liquid |
Safety and Handling
P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310-P403 + P233
missing translation for 'dotInformation' : 6.1/PGII
missing translation for 'rtecsNumber' : 203-058-7
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tributylamine, ≥98.5%, Honeywell™