Learn More
BRAND™ Triangular Wedge-Shape PTFE Magnetic Stirring Bars
An excellent option for liquid agitation in the laboratory.
Brand: BRAND™ 137820
This item is not returnable.
View return policy
Description
- Ideal for dissolving solids and mixing of sediments due to scraping action
- Angular surface creates high turbulence even at low speeds
- Provided in packs of 10
Specifications
Triangular wedge-shaped | |
50 mm | |
10/Pk. |
12 mm | |
PTFE | |
Triangle |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
51Hours
20Minutes
48Seconds
44
Product Content Correction
Product Title
BRAND™ Triangular Wedge-Shape PTFE Magnetic Stirring Bars > Diameter (Metric): 12 mm, Length (Metric): 50 mm