missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triacetin, 99%, MP Biomedicals™
Click to view available options
Quantity:
0.5 L
1 L
Specifications
Color | Colorless |
Density | 1.159g/cm3 at 20°C |
Assay Percent Range | 99% |
Quantity | 1 L |
Formula Weight | 218.2 |
Physical Form | Semi-viscous Liquid |
Chemical Name or Material | Triacetin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'