missing translation for 'onlineSavingsMsg'
Learn More

Triacetin, 99%, MP Biomedicals™

Product Code. 11473491
Click to view available options
Quantity:
0.5 L
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.205
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
Synonym triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
PubChem CID 5541
ChEBI CHEBI:9661
IUPAC Name 2,3-diacetyloxypropyl acetate
SMILES CC(=O)OCC(COC(=O)C)OC(=O)C

Specifications

Color Colorless
Density 1.159g/cm3 at 20°C
Assay Percent Range 99%
Quantity 0.5 L
Formula Weight 218.2
Physical Form Semi-viscous Liquid
Chemical Name or Material Triacetin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'