Learn More
uvex Trend BOA™ Black/Gray/Red PU/PU 69082 Low Shoe - Width 11
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1290.00 NOK
Specifications
Color | Black, Gray, Red |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17203301
|
uvex
6908238 |
38 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17213301
|
uvex
6908239 |
39 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17223301
|
uvex
6908240 |
40 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17233301
|
uvex
6908241 |
41 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17243301
|
uvex
6908242 |
42 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17253301
|
uvex
6908243 |
43 |
1290.00 NOK
1 pair |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||||
17263301
|
uvex
6908244 |
44 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17273301
|
uvex
6908245 |
45 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17283301
|
uvex
6908246 |
46 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17293301
|
uvex
6908247 |
47 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17203311
|
uvex
6908248 |
48 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17213311
|
uvex
6908249 |
49 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17223311
|
uvex
6908250 |
50 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17233311
|
uvex
6908251 |
51 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
17243311
|
uvex
6908252 |
52 |
1290.00 NOK
1 pair |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
Description
Uvex 2 Trend Perforated Shoe with BOA™ Fit System 6908.2 S1 P SRC
- Sporty, S1 P perforated safety shoe with wide fit
- Breathable perforated microvelour
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- BOA™ Fit System Dial for custom comfort that's consistent all day
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel toe cap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable climatic insole with a moisture transport system
- Helps maintain foot temperature for optimum wearer comfort
- Soft padding on collar and tongue
- Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
- Article No. 6908.2
- Standard EN ISO 20345:2011 S1 P SRC
- Sizes: 38 to 52
Specifications
Black, Gray, Red | |
Dual-density polyurethane (PU/PU) | |
Reusable | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Standard Width | |
Yes | |
S1 | |
ESD Safety Shoe |
Product Title
uvex Trend BOA™ Black/Gray/Red PU/PU 69082 Low Shoe - Width 11